accessibility menu, dialog, popup

UserName

OHBHOO

CAS RN 87199-16-4

IUPAC Name: (3-formylphenyl)boronic acid
Synonyms: 3-formylphenylboronic acid
Molecular Weight (g/mol): 149.94
Molecular Formula: C7H7BO3
InChi Key: HJBGZJMKTOMQRR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(C=O)=C1
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)