accessibility menu, dialog, popup

UserName

HOBOHO

CAS RN 87199-17-5

IUPAC Name: (4-formylphenyl)boronic acid
Synonyms: 4-formylphenylboronic acid
Molecular Weight (g/mol): 149.94
Molecular Formula: C7H7BO3
InChi Key: VXWBQOJISHAKKM-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=O)C=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results