CAS RN 872-49-1
CAS RN 872-49-1
IUPAC Name:
5-chloro-1-methyl-1H-imidazole
Synonyms:
5-chloro-1-methylimidazole
Molecular Weight (g/mol):
116.55
Molecular Formula:
C4H5ClN2
InChi Key:
NYDGOZPYEABERA-UHFFFAOYSA-N
SMILES:
CN1C=NC=C1Cl
1
–
2
of
2
results
1
5-Chloro-1-methylimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Chloro-1-methylimidazole, 98%, Thermo Scientific™
CAS: 872-49-1 Molecular Formula: C4H5ClN2 Molecular Weight (g/mol): 116.548 MDL Number: MFCD00014505 InChI Key: NYDGOZPYEABERA-UHFFFAOYSA-N Synonym: 5-chloro-1-methyl-1h-imidazole,1-methyl-5-chloroimidazole,1h-imidazole, 5-chloro-1-methyl,5-chloro-1-methylimidazole,zlchem 661,pubchem7612,acmc-209qj7,5-chloro-1-methyl imidazole,5-chloro-1-methyl-imidazole,ksc494e7d PubChem CID: 70105 IUPAC Name: 5-chloro-1-methylimidazole SMILES: CN1C=NC=C1Cl