accessibility menu, dialog, popup

UserName

CAS RN 872-53-7

O

CAS RN 872-53-7

IUPAC Name: cyclopentanecarbaldehyde
Synonyms: cyclopentanecarboxaldehyde
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: VELDYOPRLMJFIK-UHFFFAOYSA-N
SMILES: O=CC1CCCC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)