accessibility menu, dialog, popup

UserName

H3CS(R)NH3OH

CAS RN 87206-44-8

IUPAC Name: (2R)-1-hydroxy-4-(methylsulfanyl)butan-2-aminium
Synonyms: (2R)-1-hydroxy-4-(methylsulfanyl)butan-2-aminium
Molecular Weight (g/mol): 136.23
Molecular Formula: C5H14NOS
InChi Key: MIQJGZAEWQQAPN-RXMQYKEDSA-O
SMILES: CSCC[C@@H]([NH3+])CO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)