CAS RN 87239-81-4
IUPAC Name:
1-{[(propan-2-yloxy)carbonyl]oxy}ethyl (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms:
cefpodoxime proxetil
Molecular Weight (g/mol):
557.59
Molecular Formula:
C21H27N5O9S2
InChi Key:
LTINZAODLRIQIX-FBXRGJNPSA-N
SMILES:
COCC1=C(N2[C@H](SC1)[C@H](NC(=O)C(=N/OC)\C1=CSC(N)=N1)C2=O)C(=O)OC(C)OC(=O)OC(C)C
1
–
2
of
2
results
2
Cefpodoxime Proxetil, TRC
CAS: 87239-81-4 Molecular Formula: C21 H27 N5 O9 S2 Molecular Weight (g/mol): 557.6 Synonym: Cefpodoxime proxetil,(1RS)-1-[[(1-Methylethoxy)carbonyl]oxy]ethyl (6R,7R)-7-[[(2Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate IUPAC Name: 1-propan-2-yloxycarbonyloxyethyl (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(methoxymethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate SMILES: COCC1=C(N2[C@H](SC1)[C@H](NC(=O)\C(=N/OC)\c3csc(N)n3)C2=O)C(=O)OC(C)OC(=O)OC(C)C