CAS RN 872573-93-8
IUPAC Name:
5-[(quinolin-6-yl)methylidene]-2-{[(thiophen-2-yl)methyl]amino}-4,5-dihydro-1,3-thiazol-4-one
Synonyms:
5-(quinolin-6-ylmethylidene)-2-[(thiophen-2-ylmethyl)amino]-1,3-thiazol-4-one
Molecular Weight (g/mol):
351.44
Molecular Formula:
C18H13N3OS2
InChi Key:
XOLMRFUGOINFDQ-UHFFFAOYSA-N
SMILES:
O=C1N=C(NCC2=CC=CS2)SC1=CC1=CC2=CC=CN=C2C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ Ro 3306
CAS: 872573-93-8 Molecular Formula: C18H13N3OS2
2
Ro-3306, MedChemExpress
MedChemExpress Ro-3306 is a potent and selective inhibitor of CDK1, with Kis of 20 nM, 35 nM and 340 nM for CDK1, CDK1/cyclin B1 and CDK2/cyclin E, respectively.