accessibility menu, dialog, popup

UserName

ClMgBr

CAS RN 873-77-8

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 215.76
Molecular Formula: C6H4BrClMg
InChi Key:
SMILES: [Mg++].[Br-].Clc1cc[cH-]cc1
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results