CAS RN 87304-79-8
CAS RN 87304-79-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fructose-isoleucine (mixture of diastereomers), TRC
CAS: 87304-79-8 Molecular Formula: C12H23NO7 Molecular Weight (g/mol): 293.31 Synonym: N-(1-Deoxy-β-D-fructopyranos-1-yl)-L-isoleucine,[S-(R*,R*)]-1-[(1-Carboxy-2-methylbutyl)amino]-1-deoxy-β-D-fructopyranose IUPAC Name: (((3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydro-2H-pyran-2-yl)methyl)-L-isoleucine SMILES: O[C@H]1[C@H](O)[C@H](O)COC1(CN[C@H](C(O)=O)[C@@H](C)CC)O