accessibility menu, dialog, popup

UserName

H3CFFOH

CAS RN 87327-65-9

IUPAC Name: 1-(2,6-difluorophenyl)ethan-1-ol
Synonyms: 1-(2,6-difluorophenyl)ethanol
Molecular Weight (g/mol): 158.15
Molecular Formula: C8H8F2O
InChi Key: SIYWDKQSSDBLOA-UHFFFAOYNA-N
SMILES: CC(O)C1=C(F)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results