accessibility menu, dialog, popup

UserName

BFNH2HOOH

CAS RN 873566-75-7

IUPAC Name: (3-amino-4-fluorophenyl)boronic acid
Synonyms: 3-amino-4-fluorophenylboronic acid
Molecular Weight (g/mol): 154.94
Molecular Formula: C6H7BFNO2
InChi Key: SYBMNJPUZMUPGQ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results