CAS RN 87368-00-1
CAS RN 87368-00-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydroxy Gliclazide, TRC
CAS: 87368-00-1 Molecular Formula: C15 H21 N3 O4 S Molecular Weight (g/mol): 339.41 Synonym: Hydroxygliclazide,Cyclopenta[c]pyrrole, benzenesulfonamide deriv.,N-[[(Hexahydrocyclopenta[c]pyrrol-2(1H)-yl)amino]carbonyl]-4-(hydroxymethyl)benzenesulfonamide,Benzenesulfonamide, N-[[(hexahydrocyclopenta[c]pyrrol-2(1H)-yl)amino]carbonyl]-4-(hydroxymethyl)- IUPAC Name: 1-(3,3a,4,5,6,6a-hexahydro-1H-cyclopenta[c]pyrrol-2-yl)-3-[4-(hydroxymethyl)phenyl]sulfonylurea SMILES: OCc1ccc(cc1)S(=O)(=O)NC(=O)NN2CC3CCCC3C2