CAS RN 87375-91-5
CAS RN 87375-91-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dehydronitrosonisoldipine, TRC
CAS: 87375-91-5 Molecular Formula: C20H22N2O5 Molecular Weight (g/mol): 370.4 Synonym: 2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl 5-(2-Methylpropyl) Ester,2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid Methyl 2-Methylpropyl Ester,5-Methyl 3-(2-Methylprop-3-yl) 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate,Nisoldipine Impurity D SMILES: COC(=O)c1c(C)nc(C)c(C(=O)OCC(C)C)c1c2ccccc2N=O