accessibility menu, dialog, popup

UserName

OOOH

CAS RN 87392-05-0

IUPAC Name: oxolane-2-carboxylic acid
Synonyms: tetrahydro-2-furoic acid
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: UJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)