accessibility menu, dialog, popup

UserName

NOOHFFF

CAS RN 87407-12-3

IUPAC Name: 3-(trifluoromethyl)pyridine-2-carboxylic acid
Synonyms: 3-(trifluoromethyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 191.11
Molecular Formula: C7H4F3NO2
InChi Key: DMWLBOPLZYJGPT-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results