accessibility menu, dialog, popup

UserName

CAS RN 874219-27-9

BFONCH3H3CHOOH

CAS RN 874219-27-9

IUPAC Name: [3-(dimethylcarbamoyl)-4-fluorophenyl]boronic acid
Synonyms: 3-(dimethylcarbamoyl)-4-fluorophenylboronic acid
Molecular Weight (g/mol): 211
Molecular Formula: C9H11BFNO3
InChi Key: CRXFXVKQLCHKSK-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results