accessibility menu, dialog, popup

UserName

BFONH2HOOH

CAS RN 874219-34-8

IUPAC Name: (3-carbamoyl-4-fluorophenyl)boronic acid
Synonyms: 3-carbamoyl-4-fluorophenylboronic acid
Molecular Weight (g/mol): 182.95
Molecular Formula: C7H7BFNO3
InChi Key: RNNYXSWJFLHIRC-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results