CAS RN 874285-03-7
IUPAC Name:
2-(3-bromo-2-fluorophenyl)acetonitrile
Synonyms:
2-(3-bromo-2-fluorophenyl)acetonitrile
Molecular Weight (g/mol):
214.04
Molecular Formula:
C8H5BrFN
InChi Key:
JKRBRQNAVKGEHZ-UHFFFAOYSA-N
SMILES:
FC1=C(CC#N)C=CC=C1Br
1
–
1
of
1
results
1
3-Bromo-2-fluorophenylacetonitrile, 98%, Thermo Scientific Chemicals
CAS: 874285-03-7 Molecular Formula: C8H5BrFN Molecular Weight (g/mol): 214.04 MDL Number: MFCD09864662 InChI Key: JKRBRQNAVKGEHZ-UHFFFAOYSA-N Synonym: 3-bromo-2-fluorophenylacetonitrile,3-bromo-2-fluorophenyl acetonitrile,2-3-bromo-2-fluorophenyl acetonitrile,acmc-20a3z9,benzeneacetonitrile,3-bromo-2-fluoro,3-bromo-2-fluoro-phenyl-acetonitrile,2-3-bromo-2-fluorophenyl ethanenitrile PubChem CID: 26985257 IUPAC Name: 2-(3-bromo-2-fluorophenyl)acetonitrile SMILES: FC1=C(CC#N)C=CC=C1Br