accessibility menu, dialog, popup

UserName

CAS RN 874832-10-7

CH3NFFFNCO

CAS RN 874832-10-7

IUPAC Name: 3-isocyanato-2-methyl-6-(trifluoromethyl)pyridine
Synonyms: 3-isocyanato-2-methyl-6-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 202.14
Molecular Formula: C8H5F3N2O
InChi Key: BRXUOLWIERBAPA-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(=N1)C(F)(F)F)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results