accessibility menu, dialog, popup

UserName

CAS RN 874882-72-1

CH3CH3H3CNHHNNOHOOHOOH(E/Z)

CAS RN 874882-72-1

IUPAC Name: 4-[3-(tert-butylamino)-2-hydroxypropyl]-1H-indole-2-carbonitrile; but-2-enedioic acid
Synonyms: 4-[3-(tert-butylamino)-2-hydroxypropyl]-1H-indole-2-carbonitrile; butenedioic acid
Molecular Weight (g/mol): 387.44
Molecular Formula: C20H25N3O5
InChi Key: IOZROHWNOAFEMY-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CC(C)(C)NCC(O)CC1=C2C=C(NC2=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results