CAS RN 87539-45-5
CAS RN 87539-45-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6Alpha-Hydroxy-21-desacetyl Deflazacort, TRC
CAS: 87539-45-5 Molecular Formula: C23 H29 N O6 Molecular Weight (g/mol): 415.48 Synonym: 5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 6,11,21-trihydroxy-2'-methyl-, (6α,11β,16β)- (9CI),2H-Naphth[2',1':4,5]indeno[1,2-d]oxazole, 5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione deriv.,(6α,11β,16β)-6,11,21-Trihydroxy-2'-methyl-5'H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione,6α-Hydroxy-21-desacetyl Deflazacort,6α-Hydroxy-21-desacetyldeflazacort SMILES: CC1=N[C@@]2([C@@H](C[C@H]3[C@@H]4C[C@H](O)C5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]23C)O1)C(=O)CO