CAS RN 875467-41-7
CAS RN 875467-41-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
6,7-Dichloro-3,5-dihydro-1-hydroxyimidazo[1,2-a]quinazolin-2(1H)-one (Anagrelide Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6,7-Dichloro-3,5-dihydro-1-hydroxyimidazo[1,2-a]quinazolin-2(1H)-one (Anagrelide Impurity), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.