CAS RN 87553-74-0
IUPAC Name:
tert-butyl 2-amino-3-(4-hydroxyphenyl)propanoate
Synonyms:
tert-butyl 2-amino-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol):
237.3
Molecular Formula:
C13H19NO3
InChi Key:
DIGHFXIWRPMGSA-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)C(N)CC1=CC=C(O)C=C1
1
–
1
of
1
results
1
D-Tyrosine tert-butyl ester, 98%
CAS: 87553-74-0 Molecular Formula: C13H19NO3 Molecular Weight (g/mol): 237.30 MDL Number: MFCD00191035 InChI Key: DIGHFXIWRPMGSA-LDGXTIHJNA-N Synonym: h-d-tyr-otbu,tert-butyl 2r-2-amino-3-4-hydroxyphenyl propanoate,d-tyrosine,1,1-dimethylethyl ester,d-tyrosine tert-butyl ester,d-tyrosine tert.butyl ester,d-tyrosine-t-butylester,r-tert-butyl 2-amino-3-4-hydroxyphenyl propanoate,r-2-amino-3-4-hydroxy-phenyl-propionic acid tert-butyl ester PubChem CID: 7408333 IUPAC Name: tert-butyl (2R)-2-amino-3-(4-hydroxyphenyl)propanoate SMILES: CC(C)(C)OC(=O)[C@H](N)CC1=CC=C(O)C=C1