CAS RN 875634-01-8
IUPAC Name:
5-[3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]-N,N-dimethylpyridine-3-carboxamide
Synonyms:
5-[3-(2-methoxyphenyl)-1H-pyrrolo[2,3-b]pyridin-5-yl]-N,N-dimethylpyridine-3-carboxamide
Molecular Weight (g/mol):
372.43
Molecular Formula:
C22H20N4O2
InChi Key:
GYQRHHQPEMOLKH-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1C1=CNC2=NC=C(C=C12)C1=CC(=CN=C1)C(=O)N(C)C