accessibility menu, dialog, popup

UserName

CAS RN 875664-44-1

H3COBrNH2F

CAS RN 875664-44-1

IUPAC Name: 3-bromo-5-fluoro-4-methoxyaniline
Synonyms: 3-bromo-5-fluoro-4-methoxyaniline
Molecular Weight (g/mol): 220.04
Molecular Formula: C7H7BrFNO
InChi Key: DPALOVXCAGMEBU-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(N)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results