CAS RN 87585-03-3
CAS RN 87585-03-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
13-Ethyl-6alpha,17-dihydroxy-18,19-dinor-17alpha-pregn-4-en-20-yn-3-one (6alpha-Hydroxylevonorgestrel), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
6a-Hydroxy Levonorgestrel, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.