accessibility menu, dialog, popup

UserName

HClH3COONH2

CAS RN 876-33-5

IUPAC Name: O-[(4-methoxyphenyl)methyl]hydroxylamine hydrochloride
Synonyms: O-[(4-methoxyphenyl)methyl]hydroxylamine hydrochloride
Molecular Weight (g/mol): 189.64
Molecular Formula: C8H12ClNO2
InChi Key: DHEZQYZJFCIQQA-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(CON)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results