accessibility menu, dialog, popup

UserName

CAS RN 876-53-9

BrBr

CAS RN 876-53-9

IUPAC Name: 1,3-dibromoadamantane
Synonyms: 1,3-dibromoadamantane
Molecular Weight (g/mol): 294.03
Molecular Formula: C10H14Br2
InChi Key: HLWZKLMEOVIWRK-UHFFFAOYSA-N
SMILES: BrC12CC3CC(C1)CC(Br)(C3)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results