accessibility menu, dialog, popup

UserName

CAS RN 876316-31-3

NNNI

CAS RN 876316-31-3

IUPAC Name: 1-[(2-iodophenyl)methyl]-1H-1,2,4-triazole
Synonyms: 1-[(2-iodophenyl)methyl]-1,2,4-triazole
Molecular Weight (g/mol): 285.09
Molecular Formula: C9H8IN3
InChi Key: NDJIYFFWZVACMJ-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results