CAS RN 876316-33-5
CAS RN 876316-33-5
IUPAC Name:
4-{[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Synonyms:
4-{[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}morpholine
Molecular Weight (g/mol):
303.21
Molecular Formula:
C17H26BNO3
InChi Key:
WCNXVDLZTGODQW-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=CC=C1CN1CCOCC1