CAS RN 876708-03-1
CAS RN 876708-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
RO-9187, MedChemExpress
MedChemExpress RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.