accessibility menu, dialog, popup

UserName

CAS RN 876718-01-3

NNNOOH

CAS RN 876718-01-3

IUPAC Name: 2-[(1H-1,2,4-triazol-1-yl)methyl]benzoic acid
Synonyms: 2-(1,2,4-triazol-1-ylmethyl)benzoic acid
Molecular Weight (g/mol): 203.2
Molecular Formula: C10H9N3O2
InChi Key: KPKNLIVZZRDKGI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1CN1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results