CAS RN 87691-87-0
IUPAC Name:
3-(piperazin-1-yl)-1,2-benzothiazole
Synonyms:
3-(piperazin-1-yl)-1,2-benzothiazole
Molecular Weight (g/mol):
219.31
Molecular Formula:
C11H13N3S
InChi Key:
KRDOFMHJLWKXIU-UHFFFAOYSA-N
SMILES:
C1CN(CCN1)C1=NSC2=CC=CC=C12
1
–
2
of
2
results
1
3-(1-Piperazinyl)-1,2-benzisothiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(1-Piperazinyl)-1,2-benzisothiazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.