accessibility menu, dialog, popup

UserName

CAS RN 87691-89-2

NHNNO

CAS RN 87691-89-2

IUPAC Name: 3-(piperazin-1-yl)-1,2-benzoxazole
Synonyms: 3-(piperazin-1-yl)-1,2-benzoxazole
Molecular Weight (g/mol): 203.25
Molecular Formula: C11H13N3O
InChi Key: ZDFQBFVFCPABKQ-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=NOC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)