CAS RN 877-42-9
CAS RN 877-42-9
IUPAC Name:
6-bromo-2-methylquinoline
Synonyms:
6-bromo-2-methylquinoline
Molecular Weight (g/mol):
222.09
Molecular Formula:
C10H8BrN
InChi Key:
SQRYQSKJZVQJAY-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C=C(Br)C=CC2=N1
1
–
1
of
1
results
1
6-Bromo-2-methylquinoline, 97%
CAS: 877-42-9 Molecular Formula: C10H8BrN Molecular Weight (g/mol): 222.085 MDL Number: MFCD00079724 InChI Key: SQRYQSKJZVQJAY-UHFFFAOYSA-N Synonym: 6-bromoquinaldine,2-methyl-6-bromoquinoline,6-bromo-2-methyl-quinoline,quinoline, 6-bromo-2-methyl,6-bromo-2-methyl-1-azanaphthalene,zlchem 926,pubchem5882,acmc-209qps,ksc494e7l PubChem CID: 522885 IUPAC Name: 6-bromo-2-methylquinoline SMILES: CC1=NC2=C(C=C1)C=C(C=C2)Br