CAS RN 877-44-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
1,2,4-Triethylbenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
PIANO Aromatics Mixture 91, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
3
1,2,4-Triethylbenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.