accessibility menu, dialog, popup

UserName

CAS RN 877-95-2

H3COHN

CAS RN 877-95-2

IUPAC Name: N-(2-phenylethyl)acetamide
Synonyms: N-(2-phenylethyl)acetamide
Molecular Weight (g/mol): 163.22
Molecular Formula: C10H13NO
InChi Key: MODKMHXGCGKTLE-UHFFFAOYSA-N
SMILES: CC(=O)NCCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)