CAS RN 877052-79-4
CAS RN 877052-79-4
IUPAC Name:
4-{[3,5-bis(trifluoromethyl)phenyl]methyl}-6-(2-fluorophenyl)-2H,3H,4H,5H-pyrido[3,2-f][1,4]oxazepin-3-one
Synonyms:
4-{[3,5-bis(trifluoromethyl)phenyl]methyl}-6-(2-fluorophenyl)-2H,5H-pyrido[3,2-f][1,4]oxazepin-3-one
Molecular Weight (g/mol):
484.37
Molecular Formula:
C23H15F7N2O2
InChi Key:
ZIXNJVGTAXRKAP-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1C1=C2CN(CC3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(=O)COC2=NC=C1
1
–
1
of
1
results
1