CAS RN 877140-09-5
CAS RN 877140-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(6,7-Dimethoxy-4-Oxoquinazolin-3(4h)-Yl)acetic Acid, TRC
CAS: 877140-09-5 Molecular Formula: C12H12N2O5 Molecular Weight (g/mol): 264.23 Synonym: 6,7-Dimethoxy-4-oxo-3(4H)-quinazolineacetic Acid,(6,7-Dimethoxy-4-oxoquinazolin-3(4H)-yl)acetic Acid,2-(6,7-Dimethoxy-4-oxo-3,4-dihydroquinazolin-3-yl)acetic Acid,2-(6,7-Dimethoxy-4-oxoquinazolin-3-yl)acetic Acid SMILES: COc1cc2N=CN(CC(=O)O)C(=O)c2cc1OC