CAS RN 877402-45-4
CAS RN 877402-45-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
3-(Trifluoromethyl)-6,7-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-8(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-(Trifluoromethyl)-6,7-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-8(5H)-one, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
3-(Trifluoromethyl)-6,7-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-8(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
3-(Trifluoromethyl)-6,7-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-8(5H)-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.