accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNN

CAS RN 878194-92-4

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carbonitrile
Molecular Weight (g/mol): 230.07
Molecular Formula: C12H15BN2O2
InChi Key: LUDBQHAQFBWGNI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(C=NC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results