CAS RN 878489-28-2
CAS RN 878489-28-2
IUPAC Name:
2-(4-chlorophenoxy)-N-(5-methanesulfonamidoadamantan-2-yl)-2-methylpropanamide
Synonyms:
2-(4-chlorophenoxy)-N-(5-methanesulfonamidoadamantan-2-yl)-2-methylpropanamide
Molecular Weight (g/mol):
440.98
Molecular Formula:
C21H29ClN2O4S
InChi Key:
OSGIRCJRKSAODN-UHFFFAOYNA-N
SMILES:
CC(C)(OC1=CC=C(Cl)C=C1)C(=O)NC1C2CC3CC1CC(C3)(C2)NS(C)(=O)=O
1
–
1
of
1
results
1