CAS RN 878557-19-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TopFluor Cholesterol, TRC
CAS: 878557-19-8 Molecular Formula: C36H51BF2N2O Molecular Weight (g/mol): 576.61 Synonym: (T-4)-[(3beta)-24-(3,5-dimethyl-1H-pyrrol-2-yl-kN)-24-(3,5-dimethyl-2H-pyrrol-2-ylidene-kN)chol-5-en-3-olato]difluoroboron,Bodipy FL Cholesterol,Bodipy cholesterol SMILES: [F-][B+3]1([F-])[N]2=C(C)C=C(C)C2=C(CC[C@@H](C)[C@H]3CC[C@H]4[C@@H]5CC=C6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]34C)C7=C(C)C=C(C)[N-]17