CAS RN 878739-06-1
IUPAC Name:
3-(1-cyano-1-methylethyl)-N-{4-methyl-3-[(3-methyl-4-oxo-3,4-dihydroquinazolin-6-yl)amino]phenyl}benzamide
Synonyms:
3-(1-cyano-1-methylethyl)-N-{4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl}benzamide
Molecular Weight (g/mol):
451.53
Molecular Formula:
C27H25N5O2
InChi Key:
ZGBGPEDJXCYQPH-UHFFFAOYSA-N
SMILES:
CN1C=NC2=CC=C(NC3=C(C)C=CC(NC(=O)C4=CC(=CC=C4)C(C)(C)C#N)=C3)C=C2C1=O
1
–
3
of
3
results
1
Tocris Bioscience™ AZ 628
CAS: 878739-06-1 Molecular Formula: C27H25N5O2
2
3
AZ 628, MedChemExpress
MedChemExpress AZ 628 is a pan-Raf kinase inhibitor with IC50s of 105, 34 and 29 nM for B-Raf, B-RafV600E, and c-Raf-1, respectively.