CAS RN 878745-51-8
IUPAC Name:
methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
Synonyms:
methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate
Molecular Weight (g/mol):
206.12
Molecular Formula:
C7H5F3N2O2
InChi Key:
VBFLWQBZJODKRL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=NC(=NC=C1)C(F)(F)F
1
–
1
of
1
results
1
Methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate, 97%, Thermo Scientific Chemicals
CAS: 878745-51-8 Molecular Formula: C7H5F3N2O2 Molecular Weight (g/mol): 206.12 MDL Number: MFCD05662690 InChI Key: VBFLWQBZJODKRL-UHFFFAOYSA-N Synonym: methyl 2-trifluoromethyl pyrimidine-4-carboxylate,2-trifluoromethyl-pyrimidine-4-carboxylic acid methyl ester,methyl2-trifluoromethyl pyrimidine-4-carboxylate,methyl 2-trifluoromethyl-4-pyrimidine carboxylate,4-pyrimidinecarboxylic acid, 2-trifluoromethyl-, methyl ester,4-pyrimidinecarboxylicacid, 2-trifluoromethyl-, methyl ester,4-pyrimidinecarboxylicacid,2-trifluoromethyl-,methyl ester PubChem CID: 17750127 IUPAC Name: methyl 2-(trifluoromethyl)pyrimidine-4-carboxylate SMILES: COC(=O)C1=NC(=NC=C1)C(F)(F)F