accessibility menu, dialog, popup

UserName

CAS RN 879-05-0

FFFFFMgBr

CAS RN 879-05-0

IUPAC Name: magnesium(2+) pentafluorobenzen-1-ide bromide
Synonyms: magnesium(2+) pentafluorobenzen-1-ide bromide
Molecular Weight (g/mol): 271.27
Molecular Formula: C6BrF5Mg
InChi Key: AMQDBUIQKQUCKY-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].FC1=[C-]C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results