CAS RN 879-39-0
IUPAC Name:
1,2,3,4-tetrachloro-5-nitrobenzene
Synonyms:
1,2,3,4-tetrachloro-5-nitrobenzene
Molecular Weight (g/mol):
260.88
Molecular Formula:
C6HCl4NO2
InChi Key:
MTBYTWZDRVOMBR-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(Cl)=C(Cl)C(Cl)=C1Cl
1
–
3
of
3
results
1
1,2,3,4-Tetrachloro-5-nitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3,4-Tetrachloro-5-nitrobenzene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.