accessibility menu, dialog, popup

UserName

HNHNHNOH2OHClHCl

CAS RN 879089-64-2

IUPAC Name: 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H,4H,5H-pyrrolo[3,2-b]pyridin-5-one hydrate dihydrochloride
Synonyms: 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H,4H-pyrrolo[3,2-b]pyridin-5-one hydrate dihydrochloride
Molecular Weight (g/mol): 306.19
Molecular Formula: C12H17Cl2N3O2
InChi Key: UXDAADMPPAHNMY-UHFFFAOYSA-N
SMILES: O.Cl.Cl.O=C1NC2=C(NC=C2C2=CCNCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results