CAS RN 879093-08-0
IUPAC Name:
2,5-difluoro-4-methoxybenzaldehyde
Synonyms:
2,5-difluoro-4-methoxybenzaldehyde
Molecular Weight (g/mol):
172.13
Molecular Formula:
C8H6F2O2
InChi Key:
DCGKDDVUUMOTDH-UHFFFAOYSA-N
SMILES:
COC1=C(F)C=C(C=O)C(F)=C1
1
–
2
of
2
results
2
2,5-Difluoro-4-methoxybenzaldehyde, 98%, Thermo Scientific™
CAS: 879093-08-0 Molecular Formula: C8H6F2O2 Molecular Weight (g/mol): 172.13 MDL Number: MFCD06246882 InChI Key: DCGKDDVUUMOTDH-UHFFFAOYSA-N PubChem CID: 3856938 IUPAC Name: 2,5-difluoro-4-methoxybenzaldehyde SMILES: COC1=C(F)C=C(C=O)C(F)=C1