accessibility menu, dialog, popup

UserName

CAS RN 88050-17-3

HO(R)(S)NOOOHO

CAS RN 88050-17-3

IUPAC Name: (2S,4R)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-hydroxypyrrolidine-2-carboxylic acid
Synonyms: (2S,4R)-1-[(9H-fluoren-9-ylmethoxy)carbonyl]-4-hydroxypyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 353.37
Molecular Formula: C20H19NO5
InChi Key: GOUUPUICWUFXPM-XIKOKIGWSA-N
SMILES: O[C@@H]1C[C@H](N(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results